Cytidine-5'-triphosphate disodium salt, 97%
Cytidine-5'-triphosphate disodium salt, 97%
Cytidine-5'-triphosphate disodium salt, 97%
Thermo Scientific Chemicals

Cytidine-5'-triphosphate disodium salt, 97%

CAS: 36051-68-0 | C9H14N3Na2O14P3 | 527.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
2 g
Catalog number J62238.04
also known as J62238-04
Price (EUR)
645,00
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
645,00
Each
Chemical Identifiers
CAS36051-68-0
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC9H14N3Na2O14P3
InChI KeyNFQMDTRPCFJJND-WFIJOQBCSA-L
SMILES[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals≤10ppm (non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (unspecified)≥88.0% (dry wt. basis)
Assay (HPLC)≥96.0%
View more
Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhou, H.; Kong, S.; Pan, Y.; Zhang, Z.; Deng, L. Microwave-assisted fabrication of strontium doped apatite coating on Ti6Al4V. Mater. Sci. Eng., C 2015, 56, 174-180.
  2. Zheng, S.; Lu, J.; Li, W.; Qin, Y.; Yan, D.; Evans, D. G.; Duan, X. The 2-phenylbenzimidazole-5-sulfonate/layered double hydroxide co-intercalation composite and its luminescence response to nucleotides. J. Mater. Chem. C 2014, 2 (26), 5161-5167.