Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%
Thermo Scientific™

Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.9%

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number J62569.36
also known as J62569-36
Price (EUR)
135,15
Online Exclusive
159,00
Save 23,85 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
135,15
Online Exclusive
159,00
Save 23,85 (15%)
Each
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
DNaseNone detected
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥99.9%
Protease activityNone detected
View more
Used as a biological buffer substance in the pH range 7—9; pKa = 8.3 at 20°C; pKa = 7.82 at 37°C. Used as a biological buffer or as a component of buffer formulations such as TAE and TBE buffers. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms (pH 7.0-9.0). Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins. Used in chemical manufacturing (surface-active agents, vulcanization accelerators, and pharmaceuticals), as an emulsifying agent (cosmetics, mineral oil and paraffin wax emulsions, leather dressings, textile specialties, polishes, cleaning compounds, and soluble oils), and as an absorbent for acidic gases, biological buffer, acidimetric standard, and therapeutic alkalinizing agent. Used in buffer systems for the freezer storage of sperm, to regulate pH in the transport of live aquatic species, cell culture media, and anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Kallet RH, et al. The treatment of acidosis in acute lung injury with tris-hydroxymethyl aminomethane (THAM).Am J Respir Crit Care Med.,2000,161(1), 1149-53.
  2. Desmarais WT, et al. The 1.20 A resolution crystal structure of the aminopeptidase from Aeromonas proteolytica complexed with tris: a tale of buffer inhibition.Structure.,2002,10(8), 1063-72.