Laminin (925-933)
Thermo Scientific Chemicals

Laminin (925-933)

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J64340.MCR
also known as J64340-M^
1 mg
Catalog number J64340.MCR
also known as J64340-M^
Price (EUR)
113,00
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
113,00
Each
Chemical Identifiers
CAS110590-60-8
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized Powder
Appearance (Color)White
Assay from Supplier's CofAPeptide Content: Report Value
Mass SpecExhibits correct MW
Assay from Supplier's CofAPeptide Purity: Report Value
Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor, Inhibits experimental metastasis. find useful for product applications. And it is also used in Western Blot, Blocking Peptides, Dot Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytomety, Cell Culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C