Cefazolin sodium salt
Cefazolin sodium salt
Cefazolin sodium salt
Thermo Scientific Chemicals

Cefazolin sodium salt

Inhibits bacterial cell wall synthesis | CAS: 27164-46-1 | C14H16N8NaO4S3 | 479.504 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65274.06
also known as J65274-06
Price (EUR)
448,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
448,00
Each
Chemical Identifiers
CAS27164-46-1
IUPAC Namesodium 3-{[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl}-8-oxo-7-[2-(1H-1,2,3,4-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular FormulaC14H13N8NaO4S3
InChI KeyFLKYBGKDCCEQQM-UHFFFAOYNA-M
SMILES[Na+].CC1=NN=C(SCC2=C(N3C(SC2)C(NC(=O)CN2C=NN=N2)C3=O)C([O-])=O)S1
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.0 - 6.0
FormWhite to off-white crystalline powder
Assay (unspecified)89.1-110.1 % of Cefazolin sodium (anhydrous basis)
Water Content (Karl Fischer Titration)≤6.0%
Optical Rotation-10? to -24?
Inhibits bacterial cell wall synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only