Ceftriaxone sodium hemiheptahydrate
Ceftriaxone sodium hemiheptahydrate
Ceftriaxone sodium hemiheptahydrate
Thermo Scientific Chemicals

Ceftriaxone sodium hemiheptahydrate

CAS: 104376-79-6 | C36H46N16Na4O21S6 | 1323.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66003.MD
also known as J66003-MD
Price (EUR)
102,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
102,00
Each
Chemical Identifiers
CAS104376-79-6
IUPAC Nametetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
Molecular FormulaC36H46N16Na4O21S6
InChI KeyRGDBKADCOSIOEV-MAODNAKNSA-J
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
Assay from Suppliers CofAReport
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only