2,6-Dinitrotoluene, 97%
2,6-Dinitrotoluene, 97%
2,6-Dinitrotoluene, 97%
Thermo Scientific Chemicals

2,6-Dinitrotoluene, 97%

CAS: 606-20-2 | C7H6N2O4 | 182.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L00856.14
also known as L00856-14
Price (EUR)
76,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
76,20
Each
Chemical Identifiers
CAS606-20-2
IUPAC Name2-methyl-1,3-dinitrobenzene
Molecular FormulaC7H6N2O4
InChI KeyXTRDKALNCIHHNI-UHFFFAOYSA-N
SMILESCC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only