2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.

CAS: 375-02-0 | C4HF7O | 198.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L00875.03
also known as L00875-03
Price (EUR)
56,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
56,30
Each
Chemical Identifiers
CAS375-21-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
CommentPhysical state depends on water and ethyl hemiacetal content
Water Content (Karl Fischer Titration)≤15%
Proton NMRConforms to structure
FormCrystals or fused solid or clear liquid as melt
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only