Cyclopropyl phenyl ketone, 97%
Cyclopropyl phenyl ketone, 97%
Cyclopropyl phenyl ketone, 97%
Thermo Scientific Chemicals

Cyclopropyl phenyl ketone, 97%

CAS: 3481-02-5 | C10H10O | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L02218.14
also known as L02218-14
Price (EUR)
88,80
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
88,80
Each
Chemical Identifiers
CAS3481-02-5
IUPAC Namecyclopropyl(phenyl)methanone
Molecular FormulaC10H10O
InChI KeyPJRHFTYXYCVOSJ-UHFFFAOYSA-N
SMILESO=C(C1CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Refractive Index1.5510-1.5565 @ 20?C
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. W. J. Close. An Improved Synthesis of Cyclopropyl Phenyl Ketone and Related Substances. J. Am. Chem. Soc. 1957, 79 (6), 1455-1458.
  2. Frank J. Piehl.; Weldon G. Brown. Alkylation of Cyclopropyl Phenyl Ketone. J. Am. Chem. Soc. 1953, 75(20), 5023-5026.