3-(Trifluoromethyl)crotonic acid, 96%
3-(Trifluoromethyl)crotonic acid, 96%
3-(Trifluoromethyl)crotonic acid, 96%
Thermo Scientific Chemicals

3-(Trifluoromethyl)crotonic acid, 96%

CAS: 69056-67-3 | C5H5F3O2 | 154.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03816.06
also known as L03816-06
Price (EUR)
89,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
89,20
Each
Chemical Identifiers
CAS69056-67-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Refractive Index1.3810-1.3870 as melt
Appearance (Color)White to pale yellow or clear colorless to pale yellow
FormCrystals or powder or crystalline powder or fused solid or liquid as melt
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only