L-Arginine methyl ester dihydrochloride, 98%
L-Arginine methyl ester dihydrochloride, 98%
L-Arginine methyl ester dihydrochloride, 98%
Thermo Scientific Chemicals

L-Arginine methyl ester dihydrochloride, 98%

CAS: 26340-89-6 | C7H18Cl2N4O2 | 261.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05959.14
also known as L05959-14
Price (EUR)
145,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
145,00
Each
Chemical Identifiers
CAS26340-89-6
IUPAC Namedihydrogen methyl (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoate dichloride
Molecular FormulaC7H18Cl2N4O2
InChI KeyXQYZOBNLCUAXLF-XRIGFGBMSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Optical Rotation+21.0 ? 1.0? (c=2.5 in methanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only