Thermo Scientific Chemicals

1-Naphthylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L06696.06
5 g, Each
Thermo Scientific Chemicals

1-Naphthylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L06696.06
5 g, Each
Quantity
Catalog number: L06696.06
also known as L06696-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
132-75-2
IUPAC Name
2-(naphthalen-1-yl)acetonitrile
Molecular Formula
C12H9N
InChI Key
OQRMWUNUKVUHQO-UHFFFAOYSA-N
SMILES
N#CCC1=C2C=CC=CC2=CC=C1
Assay (GC)
≥96.0%
Melting Point (clear melt)
31.0-40.0?C
Appearance (Color)
White to pale cream to cream to yellow to brown
Refractive Index
1.6175-1.6225 @ 20?C
Form
Crystals or powder or crystalline powder or fused solid or clear liquid as melt

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water