Norbornane-2-carboxylic acid, 98%, predominantly endo isomer
Norbornane-2-carboxylic acid, 98%, predominantly endo isomer
Norbornane-2-carboxylic acid, 98%, predominantly endo isomer
Thermo Scientific Chemicals

Norbornane-2-carboxylic acid, 98%, predominantly endo isomer

CAS: 824-62-4 | C8H12O2 | 140.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08439.14
also known as L08439-14
Price (EUR)
305,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
305,00
Each
Chemical Identifiers
CAS824-62-4
IUPAC Namebicyclo[2.2.1]heptane-2-carboxylic acid
Molecular FormulaC8H12O2
InChI KeyJESWDXIHOJGWBP-UHFFFAOYNA-N
SMILESOC(=O)C1CC2CCC1C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5% (sum of endo and exo isomers)
FormCrystals or waxy solid
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Incompatible with oxidizing agents, Strong acids, Strong bases.
RUO – Research Use Only
Harold. Hart; Frank J. Chloupek. Decomposition of Diacyl Peroxides from exo- and endo-Norbornane-2-carboxylic Acids and exo- and endo-Norbornene-5-carboxylic Acids. J. Am. Chem. Soc. 1963, 85 (8), 1155-1160.