L-Aspartic acid 4-benzyl ester, 98%
L-Aspartic acid 4-benzyl ester, 98%
L-Aspartic acid 4-benzyl ester, 98%
Thermo Scientific Chemicals

L-Aspartic acid 4-benzyl ester, 98%

CAS: 2177-63-1 | C11H13NO4 | 223.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08956.14
also known as L08956-14
Price (EUR)
276,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
276,00
Each
Chemical Identifiers
CAS2177-63-1
IUPAC Name2-amino-4-(benzyloxy)-4-oxobutanoic acid
Molecular FormulaC11H13NO4
InChI KeyVGALFAWDSNRXJK-UHFFFAOYNA-N
SMILESNC(CC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)> 97.5%
FormPowder
Optical Rotation+27.5? ? 1.5? (c=1 in 1N HCl)
Appearance (Color)White
L-Aspartic acid β-benzyl ester is used in the synthesis of peptides with a 1,4-diazepine-2,5-dione ring structure and in development of block copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid β-benzyl ester is used in the synthesis of peptides with a 1,4-diazepine-2,5-dione ring structure and in development of block copolymers.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. K W Marck; C H Wildevuur; W L Sederel; A Bantjes; J Feijen. Biodegradability and tissue reaction of random copolymers of L-leucine, L-aspartic acid, and L-aspartic acid esters. Mutation Research/Fundamental and Molecular Mechanisms of Mutagenesis,1977, 11 (3), 405-422.
  2. Lei Wang, Rong Zeng; Chao Li; Renzhong Qiao; Lei Wang; Rong Zeng; Chao Li; Renzhong Qiao. Self-assembled polypeptide-block-poly(vinylpyrrolidone) as prospective drug-delivery systems. Colloids and Surfaces B: Biointerfaces. 2009, 74 (1), 284-292.