5-Acetamido-2-aminobenzoic acid, 97%
5-Acetamido-2-aminobenzoic acid, 97%
5-Acetamido-2-aminobenzoic acid, 97%
Thermo Scientific Chemicals

5-Acetamido-2-aminobenzoic acid, 97%

CAS: 50670-83-2 | C9H10N2O3 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11256.14
also known as L11256-14
Price (EUR)
128,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
128,00
Each
Chemical Identifiers
CAS50670-83-2
IUPAC Name2-amino-5-acetamidobenzoic acid
Molecular FormulaC9H10N2O3
InChI KeyGSOHXJQXAKNJES-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(N)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to grey to brown
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only