3,3'-Diaminodiphenyl sulfone, 98%
3,3'-Diaminodiphenyl sulfone, 98%
3,3'-Diaminodiphenyl sulfone, 98%
Thermo Scientific Chemicals

3,3'-Diaminodiphenyl sulfone, 98%

CAS: 599-61-1 | C12H12N2O2S | 248.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L11400.22
also known as L11400-22
Price (EUR)
98,50
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
98,50
Each
Chemical Identifiers
CAS599-61-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to orange
FormCrystals or powder or crystalline powder
Melting Point (clear melt)170.0-177.0?C
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
3,3'-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S.L. Liu.; R. Wang.;T.S. Chung.; M.L. Chng.; Y. Liu.; R.H. Vora. Effect of diamine composition on the gas transport properties in 6FDA-durene/3,3'-diaminodiphenyl sulfone copolyimides. Journal of Membrane Science. 2002, 202 (1-2), 165-176.
  2. Sharon E Ingram.; John J Ligga.; Richard A Pethric. Properties of epoxy nanoclay system based on diaminodiphenyl sulfone and diglycidyl ether of bisphenol F: influence of post cure and structure of amine and epoxy. Polymer International. 2007, 56 (8), 1029-1034.