Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%
Thermo Scientific Chemicals

Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%

CAS: 27639-98-1 | C12F24O4 | 664.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16614.06
also known as L16614-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS27639-98-1
IUPAC Name2,3,3,3-tetrafluoro-2-{1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy}propanoyl fluoride
Molecular FormulaC12F24O4
InChI KeyIIYQDKPSLAINNX-UHFFFAOYNA-N
SMILESFC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index<1.3000 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only