2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals
2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals
2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L16776.06
also known as L16776-06
5 g
Catalog number L16776.06
also known as L16776-06
Price (EUR)
187,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
187,00
Each
Chemical Identifiers
CAS374-99-2
IUPAC Name2,2,3,3,4,4,4-heptafluorobutan-1-aminium
Molecular FormulaC4H5F7N
InChI KeyWBGBQSRNXPVFDB-UHFFFAOYSA-O
SMILES[NH3+]CC(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Erwin Klingsberg. Synthesis of Carboxylic Acid Hydrazides and s-Triazoles of the Anthraquinone Series. J. Org. Chem. 1958, 80, (21), 5786-5789.
  2. Burton S. Marks.; George C. Schweiker. Fluorine-containing Secondary Diamines1,2. J. Am. Chem. Soc. 1958, 80, (21), 5789-5792.