2,4-Dichloro-6-iodoaniline, 97%
2,4-Dichloro-6-iodoaniline, 97%
2,4-Dichloro-6-iodoaniline, 97%
Thermo Scientific Chemicals

2,4-Dichloro-6-iodoaniline, 97%

CAS: 697-90-5 | C6H4Cl2IN | 287.909 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17659.06
also known as L17659-06
Price (EUR)
49,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
49,90
Each
Chemical Identifiers
CAS697-90-5
IUPAC Name2,4-dichloro-6-iodoaniline
Molecular FormulaC6H4Cl2IN
InChI KeyYJRSMJTVXWBFJJ-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(Cl)C=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark brown
Assay (GC)≥96.0%
FormPowder or lumps
Melting Point (clear melt)79.0-85.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only