3-Bromo-4-chlorobenzoic acid, 97%
3-Bromo-4-chlorobenzoic acid, 97%
3-Bromo-4-chlorobenzoic acid, 97%
Thermo Scientific Chemicals

3-Bromo-4-chlorobenzoic acid, 97%

CAS: 42860-10-6 | C7H4BrClO2 | 235.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19244.03
also known as L19244-03
Price (EUR)
60,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
60,60
Each
Chemical Identifiers
CAS42860-10-6
IUPAC Name3-bromo-4-chlorobenzoic acid
Molecular FormulaC7H4BrClO2
InChI KeyNLEPZGNUPNMRGF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Br)=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)215.5-224.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only