2,4-Dibromobenzenesulfonyl chloride, 98%
2,4-Dibromobenzenesulfonyl chloride, 98%
2,4-Dibromobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2,4-Dibromobenzenesulfonyl chloride, 98%

CAS: 72256-95-2 | C6H3Br2ClO2S | 334.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19887.03
also known as L19887-03
Price (EUR)
37,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
37,40
Each
Chemical Identifiers
CAS72256-95-2
IUPAC Name2,4-dibromobenzene-1-sulfonyl chloride
Molecular FormulaC6H3Br2ClO2S
InChI KeyOMOXIKAWVFDFNX-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=C(Br)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting Point (clear melt)81.5-87.5?C
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
Benjamin P. Fauber; Adam R.Johnson; Stephen Bowerman; Brenda Burton; Adam Colebrook; Annerley Flynn; Gareth Harrold; Steve Huhn; Gary Jones; Peter Lockey; Maxine Norman; Olivier Rene; Harvey Wong. Syntheses of [3H2]T0901317 and a labeled structural isomer, and characterization of the dispersed labeled compounds via 19F NMR. Journal of Labelled Compounds and Radiopharmaceuticals. 2014, 57,(1), 57-60.