4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis
4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis
4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis
Thermo Scientific Chemicals

4-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

CAS: 1736-56-7 | C9H5F3O2 | 202.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20275.06
also known as L20275-06
Price (EUR)
587,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
587,00
Each
Chemical Identifiers
CAS1736-56-7
IUPAC Name2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
Molecular FormulaC9H5F3O2
InChI KeyBGOMXTCPIUNFKR-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)C(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale pink or pale grey
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)6.27-15.13% (0.75-2.0 waters)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only