CLED Agar
Actual product may vary
Thermo Scientific™

CLED Agar

Isolate, enumerate and presumptively identify all pathogens occurring in urinary tract infections with this traditional media which provides good colonial differentiation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
PO0120A10/Pk.90mm MonoplateCLED AgarUnited Kingdom
PO5009A10/Pk.90mm MonoplateCLED AgarGermany
Catalog number PO0120A
Price (EUR)
-
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CLED Agar
Manufacturing Origin:
United Kingdom
Detect common urinary tract pathogens using Thermo Scientific™ CLED Agar, a traditional media which provides good colonial differentiation, clear diagnostic characteristics and prevents swarming from Proteus spp. due to the electrolyte deficiency in the formulation.
Isolate, enumerate and presumptively identify all UTI related microorganisms with this traditional urinary bacteriology medium which provides a valuable non-inhibitory diagnostic for plate culture.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorBlue/ Green, clear
Fill Weight19.5g ± 1.0g
Format90mm Monoplate
Media TypeCLED Agar
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life26 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 10°C