Search
Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with ready-to-use Thermo Scientific™ Oxoid™ MacConkey Agar (EP, USP, JP, BP). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water. The medium conforms to the formulations detailed in international pharmacopoeia3,4,5,6.
Use MacConkey Agar (EP, USP, JP, BP) for the cultivation of pathogens which may be present in a variety of specimens .
Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References: