ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
Thermo Scientific™

ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips

Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFormatUnit Size
3531PK
also known as 3531
200 μLSterileLift-off Lid RackPack of 960
3531200 μLSterileLift-off Lid RackCase of 100
35901000 μLNon-sterileBulkCase of 10000
3590PK
also known as 3590
1000 μLNon-sterileBulkPack of 1000
35911000 μLSterileLift-off Lid RackCase of 3200
3591PK
also known as 3591
1000 μLSterileLift-off Lid RackPack of 800
35921000 μLNon-sterileLift-off Lid RackCase of 3200
3592PK
also known as 3592
1000 μLNon-sterileLift-off Lid RackPack of 800
Catalog number 3531PK
also known as 3531
Price (HKD)
-
Volume (Metric):
200 μL
Sterility:
Sterile
Format:
Lift-off Lid Rack
Unit Size:
Pack of 960
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
  • Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
  • Ideal for genomic work
  • Universal Tip, fits most brands of pipettes
  • ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
  • Non-filtered tip
  • ART™ tips are DNase, RNase, DNA, ATP, PCR, and Endotoxin Free
  • Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
DescriptionART 200G, Non-filtered, Sterile, Racked, Lift-off Lid
DisposableSingle-use
FeaturesWide bore, sterile tip
For Use With (Application)Genomic workflows, fragile cell lines, viscous liquids
Length (English)1.96 in
Length (Metric)4.97 cm
ModelART 200G
Packaging96 tips/rack
Quantity4800/Pk.
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
MaterialVirgin Polypropylene
Product LineART
SterilitySterile
Tip StyleWide Orifice
Volume (Metric)200 μL
Unit SizePack of 960