Product Image
Thermo Scientific™
Thermo Scientific™ QSP™ Natural Screw Cap Micro Tubes
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection great with biologics, sample preparation, storage and transportation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClean ClaimsAutoclavableSterilityUnit Size
522-QPK
also known as 522-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
511-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 10
512-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileCase of 10
512-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
514-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
514-QPK
also known as 514-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
515-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileCase of 5000
515-QPK
also known as 515-Q
RNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterilePack of 500
515-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterileCase of 2500
515-S-QPK
also known as 515-S-Q
RNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterilePack of 500
516-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
516-QPK
also known as 516-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
517-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileCase of 10
517-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterileCase of 5
519-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
519-GRD-QPK
also known as 519-GRD-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
520-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
520-GRD-QPK
also known as 520-GRD-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
521-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
521-QPK
also known as 521-Q
RNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterilePack of 500
522-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 5000
522-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterileCase of 5
528-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 10
529-QRNase/DNase, DNA, pyrogen and endotoxin freeAutoclavableNon-sterileCase of 10
520-GRDS-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterileCase of 2500
529-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableSterileCase of 5
TC-512-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
TC-515-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileEach
TC-515-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
TC-517-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileEach
TC-517-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
TC-520-GRD-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileEach
TC-522-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileEach
TC-522-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
TC-529-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterileEach
TC-529-S-QRNase/DNase, DNA, pyrogen and endotoxin, ATP, PCR Inhibitor and bioburden freeNot AutoclavableNon-sterileCase of 5
Catalog number 522-QPK
also known as 522-Q
Price (HKD)
-
Clean Claims:
RNase/DNase, DNA, pyrogen and endotoxin free
Autoclavable:
Autoclavable
Sterility:
Non-sterile
Unit Size:
Pack of 500
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes.
Packaged in bulk bags and non-sterile clean claims are; RNase/DNase, DNA, pyrogen and endotoxin free.
  • Screw Cap micro tube volume range of 0.5, 1.5 and 2.0ml and are ideal for sample preparation, storage and transportation
  • Conical bottom or self standing offered with knurls in ultra clear natural or UV protective amber color
  • Tether caps and standard non tether caps are available in a rainbow of color options
Specifications
Clean ClaimsRNase/DNase, DNA, pyrogen and endotoxin free
AutoclavableAutoclavable
ColorClear
GraduatedYes
MaterialPolypropylene
SterilityNon-sterile
Unit SizePack of 500