Thermo Scientific™ QSP™ Plug Cap Tube Closure
Actual product may vary
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™

Thermo Scientific™ QSP™ Plug Cap Tube Closure

Thermo Scientific™ QSP™ Plug Caps make a snug fit, leak resistant closure for 5ml (12x75), 12mm or 13mm tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorUnit Size
540-QPK
also known as 540-Q
WhitePack of 1000
540-B-QBlueCase of 5000
540-G-QGreenCase of 5000
540-N-QNaturalCase of 5000
540-QWhiteCase of 5000
540-R-QRedCase of 5000
540-Y-QYellowCase of 5000
Catalog number 540-QPK
also known as 540-Q
Price (HKD)
-
Color:
White
Unit Size:
Pack of 1000

Thermo Scientific™ QSP™ Plug Cap Tube Closure fits 5ML, 12x75 tubes, 12mm and 13mm tubes, knurled cap helps with grip and the cap collar has ribs securing cap in the tube, Blue color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.

  • Non-autoclavable low density polyethylene material
  • Colored options
  • Plug closure leak resistant
Specifications
Clean ClaimsNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
Closure Size5 mL
For Use With (Equipment)5 mL (12x75), 12 mm or 13 mm Tubes
AutoclavableNot Autoclavable
ColorWhite
MaterialLDPE
SterilityNon-sterile
Unit SizePack of 1000