Product Image
Trek Diagnostic System

Anaerobic MIC Sensititre Plate

Perform high-quality susceptibility testing for anaerobes and B. fragilis group with the Thermo Scientific™ Sensititre™ Anaerobe MIC Plate.
Have Questions?
Catalog NumberQuantity
ANO2B10
Catalog number ANO2B
Price (HKD)
-
Quantity:
10
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on. For Research Use Only. Not for use in diagnostic procedures.
  • Recommended Applications - Healthcare
  • 5 McFarland Standard MHB (Part No. T346205)
  • Brucella Broth (Part No. T3450)
  • Description - Sensititre Anaerobe MIC Plate
  • Incubation Time - 46-48 hr
  • Incubation Conditions 34-36°C O2
  • Isolates Per Plate -1
  • Reading Options -Manual
  • Volume Per Well -100μL
  • Transfer Volume -100μL
Specifications
Certifications/ComplianceARTG
For Use With (Application)Trek Sensititre
Format10 Pc./Pk.
No. per Pack10
Packaging TypeBox
Quantity10
Shelf Life2 Years
Manufacturing OriginUnited Kingdom
Product TypeAnaerobic Plate
SpeciesB. fragilis Group
Target Organism ClassAnaerobes
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach
Contents & Storage
Ambient