GeneChip™ Fluidics Station 450Dx v.2
Applied Biosystems™

GeneChip™ Fluidics Station 450Dx v.2

The GeneChip Fluidics Station 450Dx v.2 is a component of the GeneChip System (GCS) 3000Dx v.2. The GCS 3000Dx v.2Read more
Have Questions?
Catalog NumberQuantity
00-03351 each
Catalog number 00-0335
Price (INR)
-
Quantity:
1 each

The GeneChip Fluidics Station 450Dx v.2 is a component of the GeneChip System (GCS) 3000Dx v.2. The GCS 3000Dx v.2 is the first microarray platform that is regulatory cleared for diagnostic use in the United States, Europe, and is registered in multiple other countries.

The GCS 3000Dx v.2 is a robust microarray platform for clinical applications. It is the only FDA-cleared, IVDR-compliant microarray system for RNA- and DNA-based clinical tests.

Take the proven path to market
The GCS 3000Dx v.2 consists of the GeneChip Scanner 3000Dx v.2, the GeneChip Fluidics Station 450Dx v.2, and a workstation with GeneChip Data Collection (GCDC) Dx Software. GCDC Dx Software provides an intuitive user interface that is designed to meet the needs of the diagnostic lab, ensure patient privacy, and meet the regulatory requirements for a Class II Medical Device.

The GCS 3000Dx v.2 is the foundation of the Applied Biosystems clinical toolkit, which includes cGMP-manufactured microarrays. Clinical partners can also obtain design and development support from us for the commercialization of their own cleared diagnostic products under our partnership program.

Please refer to the Documents section of this page for user guides and literature for the GCS 3000Dx v.2.

Specifications
Dimensions (L x W x H)Fluidics Station: 28 W x 16 1/8 D x 15 13/16 in. H (71.1 x 41 x 40.2 cm)
For Use With (Application)GeneChip array processing
Product LineGeneChip
Quantity1 each
Weight (English)Fluidics Station: 80 lb.
Weight (Metric)36.3 kg
Unit SizeEach

Frequently asked questions (FAQs)

I am getting an error while using the GeneChip Fluidics Station 450Dx v.2 (Cat. No. 00-0335). How can I determine which step I was on in the protocol, when the error occurred?

The following steps outline how to determine which step in the protocol you were on:
- Open Individual Station ID
- Under the steps, click the down arrow, and the step you were on should be highlighted
- After troubleshooting the error, resume at the step that was previously highlighted

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I set up the dip switch for my GeneChip Fluidics Station 450Dx v.2 (Cat. No. 00-0335)?

Please refer to pages 79-80 in the Affymetrix GeneChip Fluidics Station 450 User's Guide for AGCC (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/fs450_agcc_user_guide.pdf) for instructions on how to set up the dip switch.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I prepare my GeneChip Fluidics Station 450Dx v.2 (Cat. No. 00-0335) for long-term storage?

For long-term storage, we recommend running the shutdown process with water and running the shutdown process again with air. Remove the tubes from the wash buffer bottles and remove the peristaltic tubing. The fluidics station should now be ready for long-term storage.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.