ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set
Product Image
Invitrogen™

ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set

The ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set is a component of the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000-99).Read more
Have Questions?
Catalog NumberQuantity
00-19001
also known as 00-19001-99
1 Kit
Catalog number 00-19001
also known as 00-19001-99
Price (INR)
26,732.00
線上優惠
Ends: 31-Mar-2026
44,553.00
Save 17,821.00 (40%)
Each
Add to cart
Quantity:
1 Kit
Price (INR)
26,732.00
線上優惠
Ends: 31-Mar-2026
44,553.00
Save 17,821.00 (40%)
Each
Add to cart

The ViewRNA™ Cell Plus Fixation/Permeabilization Buffer Set is a component of the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000-99). These buffers can be used to evaluate antibody performance following the fixation and permeabilization steps of the assay. Note that additional changes to antibody performance may be observed following the hybridization steps of the ViewRNA™ Cell Plus Assay.

Please refer to the ViewRNA™ Cell Plus Assay protocol on the product data sheet for instructions for use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Kit
Product TypeFixn/Perm Buffer Set
Solution TypeFixation/Permeabilization Buffer
Unit SizeEach
Contents & Storage
• ViewRNA™ Cell Plus Fixation/Permeabilization Component A (Cat. No. 00-19500-23)
• ViewRNA™ Cell Plus Fixation/Permeabilization Component B (Cat. No, 00-19900-23)
• ViewRNA™ Cell Plus Blocking/Antibody Diluent (Cat. No. 00-19600-54)

Store all at 2°C to 8°C.