eBioscience™ Cell Stimulation Cocktail (500X)
Invitrogen™
eBioscience™ Cell Stimulation Cocktail (500X)
The Cell Stimulation Cocktail (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycinRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4970-034 x 100 μL
00-4970-93100 μL
Catalog number 00-4970-03
Price (INR)
21,849.00
線上優惠
Ends: 31-Mar-2026
33,613.00
Save 11,764.00 (35%)
Each
Quantity:
4 x 100 μL
Price (INR)
21,849.00
線上優惠
Ends: 31-Mar-2026
33,613.00
Save 11,764.00 (35%)
Each
The Cell Stimulation Cocktail (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. This cocktail can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays. It can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression.
Reported Application
Flow Cytometric Analysis, Functional Assays, Cell Culture, ELISA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity4 x 100 μL
Volume (Metric)4 x 100μL
Product LineeBioscience
Product TypeCell Stimulation Cocktail
Unit SizeEach