Product Image
Invitrogen™
eBioscience™ Fixation/Permeabilization Diluent
The Fixation/Permeabilization Diluent is intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 andRead more
Have Questions?
Catalog NumberQuantity
00-5223-56100 mL
Catalog number 00-5223-56
Price (INR)
5,234.00
Each
Quantity:
100 mL
Price (INR)
5,234.00
Each
The Fixation/Permeabilization Diluent is intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines. This must be used in combination with the Fixation/Permeabilization Concentrate (cat. 00-5123) and the Permeabilization Buffer (10X) (cat. 00-8333). Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3 Transcription Factor Buffer Staining Set (cat. 00-5523).
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 mL
Reagent TypeFixation Diluent, Permeabilization Diluent
Product LineeBioscience
Product TypeFixation/Permeabilization Diluent
Unit SizeEach