eBioscience™ Foxp3 / Transcription Factor Fixation/Permeabilization Concentrate and Diluent
Invitrogen™
eBioscience™ Foxp3 / Transcription Factor Fixation/Permeabilization Concentrate and Diluent
For staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines. Should be used with the Permeabilization Buffer. All three components sold together as the Foxp3 / Transcription Factor Staining Buffer Set.
Have Questions?
Catalog NumberQuantity
00-5521-001 kit
Catalog number 00-5521-00
Price (INR)
10,895.00
Online offer
Ends: 31-Mar-2026
16,761.00
Save 5,866.00 (35%)
Each
Quantity:
1 kit
Price (INR)
10,895.00
Online offer
Ends: 31-Mar-2026
16,761.00
Save 5,866.00 (35%)
Each
The Foxp3 Transcription Factor Fixation/Permeabilization Concentrate and Diluent solutions for staining with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines. It should be used in combination with the Permeabilization Buffer (10X) (cat. 00-8333). All three components can be purchased together in the Foxp3 / Transcription Factor Staining Buffer Set (cat. 00-5523).
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeFixation Diluent, Fixation Reagent, Permeabilization Diluent, Permeabilization Reagent
Product LineeBioscience
Product TypeFixation/Permeabilization Diluent
Unit SizeEach