Axiom™ Biobank Plus Genotyping Array
Applied Biosystems™

Axiom™ Biobank Plus Genotyping Array

The Axiom™ Biobank Plus Genotyping Arrays were designed by and for thought leaders in the human genetics community for high-throughput,Read more
Have Questions?
Catalog NumberQuantity
0008541 each
Catalog number 000854
Price (INR)
-
Quantity:
1 each
The Axiom™ Biobank Plus Genotyping Arrays were designed by and for thought leaders in the human genetics community for high-throughput, high-value genotyping of large sample cohorts to explore the genetics of complex diseases and translational research with a single comprehensive low-cost solution.

GWAS
• Intelligent marker selection enables imputation of millions of additional SNPs
• The genomic content module of each array is optimized for specific populations and is customizable to any 1000 Genomes population study by adding additional markers of your choosing

Transplant
• Content specific for transplantation research including functional variants, loss-of-function markers, and copy number
• Immune function, including content covering the major histocompatibility complex (MHC), human leukocyte antigen (HLA), and killer-cell immunoglobulin-like receptors (KIR)

Pharmacogenomic markers
• Markers were selected to represent phases of absorption, distribution, metabolism, and excretion (ADME)

Inflammation and HLA
• Contains markers with evidence for association with autoimmune and inflammation
• Covers variants in genes in the HLA and KIR regions known to be important in immune response

Exome
• Rare, non-synonymous coding SNPs and indels in protein coding regions of the genome
• A majority of variants are rare with minor allele frequency (MAF) <1%
• Newly discovered loss-of-function content
• SNPs and indels identified from a sequencing initiative of 26,000 individuals
• Known disease-causing mutationseQTLs

• Markers that have known associations to RNA expression traits

• Captures unique eQTLs contained in the NCBI Genotype-Tissue Expression (GTEx) eQTL database, GEUVIDAS project, and several other discovery projects


Human disease
• Alzheimer-s disease, including coverage of ApoE
• Blood phenotypes
• Cancer common variants
• Cardiometabolic


Choose your content
• Order the pre-designed array and customize it with markers of your choice
• Custom markers may be de novo or selected from the Applied Biosystems Axiom Genomic Database of genotype-tested markers

Please note: Reagent kits do not include plastic consumables required to run the assay on the Beckman Biomek™ FXP Target Prep Express System. Please contact us for a list of required plastic consumables.

Specifications
Quantity1 each
ArrayGenotyping
Format96-array Plate
Number of Arrays1 array
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.