Anti-Clumping Agent
Gibco™

Anti-Clumping Agent

GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densitiesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
0010057DG100 mL
0010057AE20 mL
Catalog number 0010057DG
Price (INR)
36,618.00
Each
Quantity:
100 mL
Price (INR)
36,618.00
Each
GIBCO™ Anti-Clumping Agent is a concentrated liquid reagent that reduces cell clumping to assist in attaining higher viable cell densities in suspension culture. Anti-Clumping Agent is a gentle, animal origin-free, chemically defined formulation that contains no proteins, enzymes, hydrolysates, or components of unknown composition.

• Reduced cell clumping in suspension culture
• Higher viable cell densities
• Animal origin-free and chemically defined components
• Convenient packaging with a larger size now available
• Customization available to configure packaging and delivery

Simple and Gentle Cell Dissociation
Cell lines adapted to suspension culture can revert to a more adherent or aggregated “clumping” state. When this occurs, culture condition can be greatly improved by adding a gentle reagent to reduce clumping and adhesion instead of the harsh enzymes often found in cell dissociation products. Anti-Clumping Agent is a simple, gentle, concentrated solution which, when added to culture media (usually titrated between a 1:100 and 1:1000 dilution), can reduce cell clumping significantly.

Reduced Clumping, Increased Viability, Improved Protein Expression
Reduced cell clumping achieved through use of Anti-Clumping Agent is illustrated below (Fig 1). See also the data demonstrating improved viable cell density (Fig 2). Higher viable cell density contributes to the stabilization of culture conditions for better protein yield.

For actual cell dissociation, we recommend our animal origin-free TrypLE™ products.

Use Anti-Clumping Agent after Transfection
Anti-Clumping Agent is not recommended in all suspension cell culture conditions. For example, when creating a cell line using lipid cationic transfection, Anti-Clumping Agent can interfere with the transfection process, so in this case we recommend that Anti-Clumping Agent be used after transfection.

Animal Origin-free and Chemically Defined Components
Because Anti-Clumping Agent is made with only animal origin free and chemically defined components, you can be assured that the risk of adding adventitious agents to your culture system is minimized. Avoiding additives in your system that are not well defined can lead to more robust results from batch to batch.

Convenient and Customizable Packaging
Previously, Anti-Clumping Agent was only available in a 20 mL bottle. However, it is now available in a more economical and convenient 100 mL bottle. For even larger packaging, you can custom configure this product by clicking on the 'Customize this Product' link under the 'Add to Cart' button above or by emailing us at CustomMedia@lifetech.com.

Additional Information and Support
Please review the Manuals and Protocols provided below for additional information on successful use of this product. Direct any unanswered questions to our Technical Services group.

For Research Use Only. Not for use in diagnostics procedures.
Specifications
Cell LineCHO, Insect Cells, 293 (HEK)
Cell TypeMammalian Cells
Culture TypeSuspension Cell Culture
FormatBottle(s)
Product LineGibco
Quantity100 mL
Shipping ConditionRoom Temperature
SourceAnimal origin-free
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
FormLiquid
Product TypeAnti-clumping Reagent
pH6 to 8
Unit SizeEach
Contents & Storage
Store at 2 to 8°C

Frequently asked questions (FAQs)

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.