DRI™ Amphetamine Drugs of Abuse Assays
Thermo Scientific™
DRI™ Amphetamine Drugs of Abuse Assays
Eliminate time-consuming steps while achieving high analytical accuracy when you use Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00171 x 100 mL
00181 x 500 mL
100145853 x 18 mL
Catalog number 0017
Price (INR)
-
Quantity:
1 x 100 mL
Eliminate time-consuming steps while achieving high analytical accuracy when you use Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays—ready to use in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Amphetamine Drugs of Abuse Assays
Quantity1 x 100 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesAmphetamine (AMP), Methamphetamine (mAMP)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C