Product Image
Thermo Scientific™
DRI™ Urine Drugs of Abuse Calibrators
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with these ready-to-use liquid assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
0034Low Calibrator
0036High Calibrator
Catalog number 0034
Price (INR)
-
Type:
Low Calibrator
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Urine Drugs of Abuse Calibrators—ready-to-use assays in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ConcentrationMultianalyte
Packaging1 Bottle
Quantity1 x 5 mL
TypeLow Calibrator
Detectable AnalytesAmphetamine, Cocaine, Opiate, Phencyclidine (PCP)
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C