DRI™ Benzodiazepine Drugs of Abuse Assays
Thermo Scientific™
DRI™ Benzodiazepine Drugs of Abuse Assays
Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00391 x 100 mL
00401 x 500 mL
Catalog number 0039
Price (INR)
-
Quantity:
1 x 100 mL
Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers and have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Primary DAU Control Set
DescriptionDRI Benzodiazepine Drugs of Abuse Assays
Quantity1 x 100 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesOxazepam
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C