The Thermo Scientific™ 15X Cassegrain condenser with fixed compensation for Nicolet FT-IR microscopes provides optimized measurements for windows beneath the sample.
Have Questions?
Change view
Catalog Number
Compensation
For Use With (Equipment)
0045-401
0
Nicolet Continuum, Momentum
0045-436
2 mm
Nicolet Continuum, Momentum
0047-415
0
Nicolet Infrared Microscopes
0047-417
2 mm
Nicolet Infrared Microscopes
4 Options
Catalog number 0045-401
Price (INR)
-
Compensation:
0
For Use With (Equipment):
Nicolet Continuum, Momentum
The Thermo Scientific™ 15X Cassegrain condenser with fixed compensation for Nicolet FT-IR microscopes provides optimized measurements for windows beneath the sample.
Three compensation configurations are available—zero, 1mm and 2mm. The zero compensation configuration is optimized for transmission measurements when no window material is beneath the sample. The 1mm compensation version is optimized for when a 1mm window is placed below the sample, and the 2mm compensation version is for when a 2mm window is used.
Unique design for low cost and durability
Easily purged
Recommended for:
Textiles
Forensics
Polymers
Pharmaceuticals
Specifications
Compensation0
Description15X Infinity Corrected cassegrain Condenser; Fixed compensation; Zero compensation
For Use With (Application)Spectroscopy Elemental Isotope Analysis
For Use With (Equipment)Nicolet Continuum, Momentum