Variable Compensation Condensers
Actual product may vary
Thermo Scientific™
Variable Compensation Condensers
The Thermo Scientific™ Variable Compensation Condensers correct for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
0045-40315X Infinity Corrected Condenser
0042-50710X Condenser
0045-40532X Infinity Corrected Condenser
0047-41815X Condenser with Variable Compensation
0047-41932X Condenser with Variable Compensation
Catalog number 0045-403
Price (INR)
-
Description:
15X Infinity Corrected Condenser
The Thermo Scientific™ Variable Compensation Condensers correct for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness).
Variable compensation
Easily purged
10X Reflachromat Condenser
0.71 N.A.
Standard condenser for all IRμs, Nic-Plan and IR-Plan research microscopes (not available for any other microscopes)
15X Reflachromat Condenser
0.58 N.A.
Corrects for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness)
15X Reflachromat Condenser
0.65 N.A.
Increased magnification allows it to be matched to the objective
Recommended for:
Textiles
Polymers
Pharmaceuticals
Packaging
Forensic
Specifications
Description15X Infinity Corrected Condenser
For Use With (Application)Spectroscopy Elemental Isotope Analysis
For Use With (Equipment)Nicolet Continuum, Momentum
TypeCondenser
Unit SizeEach