15X Cassegrain Objectives (Fixed Compensation)
Actual product may vary
Thermo Scientific™
15X Cassegrain Objectives (Fixed Compensation)
The Thermo Scientific™ 0.58 N.A., 15X Cassegrain objectives with fixed compensation come in three compensation configurations—zero, 1mm, or 2mm compensation objectives.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCompensationFor Use With (Equipment)
0045-4352 mmNicolet Continuum, Momentum
0045-4000Nicolet Continuum, Momentum
0047-4100All Microscopes
0047-4111 mmIRus, Nic-Plan, IR-Plan Research Microscopes
0047-4122 mmAll Microscopes
Catalog number 0045-435
Price (INR)
-
Compensation:
2 mm
For Use With (Equipment):
Nicolet Continuum, Momentum
The Thermo Scientific™ 0.58 N.A., 15X Cassegrain objectives with fixed compensation come in three compensation configurations—zero, 1mm, or 2mm compensation objectives.
The 0.58 N.A., 15X Fixed Cassegrain Objective has a working distance of 15mm. Objectives are available with three compensation configurations—zero, 1mm, and 2mm.The zero compensation objective is optimized for transmission/reflectance measurements when no window material is above the sample, the 1mm compensation objective is optimized for measurements when a 1mm thick window is above the sample, and the 2mm compensation objective is optimized for a 2mm window above the sample.
  • Prealigned objectives
  • Unique design for low cost and durability
  • Easily purged
Recommended for:
  • Forensics
  • Polymers
  • Pharmaceuticals
  • Packaging
Specifications
Compensation2 mm
Description15X Infinity Corrected Cassegrain objective; Fixed compensation; 2mm compensation
For Use With (Application)Spectroscopy Elemental Isotope Analysis
For Use With (Equipment)Nicolet Continuum, Momentum
Unit SizeEach