15X Cassegrain Condensers (Fixed Compensation)
Actual product may vary
Thermo Scientific™
15X Cassegrain Condensers (Fixed Compensation)
The Thermo Scientific™ 15X Cassegrain condenser with fixed compensation for Nicolet FT-IR microscopes provides optimized measurements for windows beneath the sample.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCompensationFor Use With (Equipment)
0047-4150Nicolet Infrared Microscopes
0045-4010Nicolet Continuum, Momentum
0045-4362 mmNicolet Continuum, Momentum
0047-4172 mmNicolet Infrared Microscopes
Catalog number 0047-415
Price (INR)
-
Compensation:
0
For Use With (Equipment):
Nicolet Infrared Microscopes
The Thermo Scientific™ 15X Cassegrain condenser with fixed compensation for Nicolet FT-IR microscopes provides optimized measurements for windows beneath the sample.
Three compensation configurations are available—zero, 1mm and 2mm. The zero compensation configuration is optimized for transmission measurements when no window material is beneath the sample. The 1mm compensation version is optimized for when a 1mm window is placed below the sample, and the 2mm compensation version is for when a 2mm window is used.
  • Unique design for low cost and durability
  • Easily purged
Recommended for:
  • Textiles
  • Forensics
  • Polymers
  • Pharmaceuticals
Specifications
Compensation0
Description15X Cassegrain Condenser; Fixed compensation; Zero compensation
For Use With (Application)Spectroscopy Elemental Isotope Analysis
For Use With (Equipment)Nicolet Infrared Microscopes
Unit SizeEach