ZipChip™ Interface
Actual product may vary
ZipChip™ Interface
ZipChip™ Interface
ZipChip™ Interface
Thermo Scientific™

ZipChip™ Interface

Use the ZipChip Interface to rapidly prepare, separate, and electrospray biological samples directly into a mass spectrometer to create a seamless CE/ESI-MS workflow.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
00950-01-00493Manual ZipChip Interface
00950-01-00492Autosampler ZipChip Interface
Catalog number 00950-01-00493
Price (INR)
-
Type:
Manual ZipChip Interface

Integrate capillary electrophoresis (CE) and electrospray ionization (ESI) into a single microfluidic device to rapidly prepare, separate, and electrospray biological samples directly into a mass spectrometer (MS). The portable size ZipChip Interface directly mounts onto select models of Thermo Scientific™ mass spectrometers, and creates a seamless CE/ESI-MS workflow that offers fast CE separation, nano-spray level sensitivity, and HRAM spectrometry for the characterization of intact proteins, antibody drug conjugates, antibody subunits, peptides, and metabolites.

Fast, sensitive, minimal sample preparation and consumption
  • Fast run times; most analyses can be done within 2-3 minutes
  • High sensitivity; stable nano-spray and nano-spray level sensitivity
  • Low (picogram to nanogram) sample consumption
  • Minimal sample preparation with online desalting
Seamless workflow for BioPharma applications

Compatible sample matrices include but not limited to:

  • Cell culture media
  • Cell lysates
  • Blood
  • Plasma
  • Urine
Mass spectrometry compatibility

Use the ZipChip Interface with Thermo Scientific™ Q Exactive™, Exactive™, Orbitrap Elite™, and LTQ Orbitrap XL™ mass spectrometers.

ZipChip autosampler*

Inject samples from vials or 96 well plates and achieve unattended 3-minute sample runs using the ZipChip Autosampler with the ZipChip Interface.

Choice of chips*
  • ZipChip HS has a shorter separation channel with the shortest run times, and can be used for small molecule, peptides, metabolites, and simple samples.
  • ZipChip HR has a longer separation channel for run times that are a little longer. The HR chip can be used for analysis of intact proteins, ADCs, antibody sub-units, complex samples, and when better resolution is required.
Assay kits*

Assay kits contain background electrolyte and sample diluent. They are convenient and reduce risk of error. Three assay kits are available:

  • Intact Protein Assay Kit
  • Peptide Mapping Assay Kit
  • Metabolite Assay Kit

*sold separately

The ZipChip Interface is a Class 1 laser product in compliance with 21 CFR 1040.10 and 1040.11 except for deviations pursuant to laser notice No. 50.

Specifications
For Use With (Equipment)Mass Spectrometers with CE/ESI
SoftwareZipChip Software
TypeManual ZipChip Interface
Unit SizeEach