ZipChip™ Interface HR and HS Chips
Actual product may vary
ZipChip™ Interface HR and HS Chips
ZipChip™ Interface HR and HS Chips
Thermo Scientific™
ZipChip™ Interface HR and HS Chips
Choose between two chips for use on your ZipChip™ Interface for Mass Spectrometry (MS). They are optimized by biological sample type.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
00950-01-00499ZipChip HR, 22cm
00950-01-00498ZipChip HS,10cm
Catalog number 00950-01-00499
Price (INR)
-
Type:
ZipChip HR, 22cm
Choose between two chips for use on your ZipChip™ Interface. The ZipChip Interface desalts, performs capillary electrophoresis (CE) separation, and nano-electrosprays a wide range of biological samples directly into Thermo Scientific™ mass spectrometer (MS) systems within a few minutes. A built-in chip recognition system allows ZipChip software to recognize the type of the chip, automatically set appropriate operational parameters and track chip usage.
Two types of chips suit for different analysis:
  • ZipChip™ HS has a shorter separation channel, and suits for small molecules, metabolites, or simple sample mixture
  • ZipChip™ HR has a longer separation channel, and suits for intact proteins, antibody drug conjugates (ADCs), antibody sub-units, or complex sample mixture
Compatible sample matrices include but not limited to:
  • Cell culture media
  • Cell lysates
  • Blood
  • Plasma
  • Urine
Fast, sensitive, convenient
  • Fast run times; most analyses can be done within 2-3 minutes
  • High sensitivity; stable nano-spray and nano-spray level sensitivity
  • Low (picogram to nanogram) sample consumption
  • Minimal to no sample preparation with online desalting
Specifications
For Use With (Equipment)ZipChip Interface
IncludesMicrofluidic Chip for Capillary Eletrohoresis, Desalting, Electospary Ionization, Sample Injection Into MS
Quantityup to 125 injections
TypeZipChip HR, 22cm
Unit SizeEach