Orion™ Conductivity Standards and Solutions
Orion™ Conductivity Standards and Solutions
Thermo Scientific™

Orion™ Conductivity Standards and Solutions

Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingConductivity
01100510Pouch111.9 mS/cm
011008Bottle100 μS/cm
011007Bottle1413 μS/cm
011005Bottles111.9 mS/cm
011006Bottle12.9 mS/cm
990106Bottle12.9 mS
01100910Pouch147 μS/cm
01100710Pouch1413 μS/cm
01100610Pouch12.9 mS/cm
Catalog number 01100510
Price (INR)
-
Request A Quote
Packaging:
Pouch
Conductivity:
111.9 mS/cm

Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.

Conductivity Standards

  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes

Warranty: Out of Box

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Boiling Point100°C
ColorClear
Conductivity111.9 mS/cm
DescriptionConductivity Standard
For Use With (Equipment)Orion pH Meter
PackagingPouch
Quantity10 x 15 mL
SolubilitySoluble
Solution TypeStandard Solution
Traceability to NISTYes
TypeConductivity Standard
pH4.75 to 7.75
Unit SizePack of 10