Product Image
Thermo Scientific™
DRI™ THC (Cannabinoid) Drugs of Abuse Assays
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
01861 x 500 mL
01851 x 100 mL
100146653 x 18 mL
Catalog number 0186
Price (INR)
-
Quantity:
1 x 500 mL
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsDRI Multi-Level THC Controls
DescriptionDRI THC (Cannabinoid) Drugs of Abuse Assays
Quantity1 x 500 mL
Sample TypeUrine
TypeReagent
Detectable Analytes11-nor-Δ9THC-COOH, 11-nor-Δ9THC-COOH
DoA CalibratorsMulti-level THC Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C