Deionization Cartridge Assembly
Product Image
Thermo Scientific™
Deionization Cartridge Assembly
For Use With: Chambers with humidity that have no DI water source
Have Questions?
Catalog NumberInstallation
019-168-00Customer Installed
Catalog number 019-168-00
Price (INR)
-
Request A Quote
Installation:
Customer Installed
Use this Thermo Scientific™ Deionization Cartridge Assembly in chambers with humidity that have no DI water source.
Specifications
DescriptionDeionization Cartridge Assembly
For Use With (Equipment)Chambers with humidity that have no DI water source
IncludesDeionization cartridge, wall bracket, pressure regulator and gauge and 1/4 in. flexible tubing
InstallationCustomer Installed
LightingLED
TypeDeionization Cartridge Assembly
Unit SizeEach