You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
pH and Electrochemistry
▸
Probes and Electrodes
▸
Dissolved Oxygen Probe Accessories
▸
Eutech™ X5 Membranes and O-rings for Dissolved Oxygen Probes
Thermo Scientific™
Eutech™ X5 Membranes and O-rings for Dissolved Oxygen Probes
Optimize your Eutech™ DO probes with these accessories and replacement parts.
Have Questions?
Catalog Number
Type
01X241603
Membrane and O-Ring
Catalog number
01X241603
Price (INR)
-
Request A Quote
Type:
Membrane and O-Ring
Optimize your Eutech™ DO probes with these accessories and replacement parts.
Membrane/O-ring Replacement is an optional procedure
It is recommended only experienced service personnel can perform this procedure.
It should only be performed if you have new membrane and O-ring.
Specifications
Description
Membrane & O-ring (pack of 5)
Probe Type
DO
Quantity
5 Pk.
For Use With (Equipment)
DO Probe
Type
Membrane and O-Ring
Unit Size
Each