Eutech™ Replacement Electrode Arm with Bracket for Benchtop Meters
Thermo Scientific™
Eutech™ Replacement Electrode Arm with Bracket for Benchtop Meters
Eutech™ Electrode Arm with bracket allows convenient hands-free operation.
Have Questions?
Catalog number 01X321801
Price (INR)
-
Eutech™ Replacement Electrode Arm with Bracket for Benchtop Meters allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
Features:
  • Swivel arm allows you to move electrode side to side or up and down while keeping electrode at a constant vertical angle.
  • Used With: CyberScan 6000, 700 and 2700 series meters
Specifications
DescriptionElectrode Arm Replacement with Bracket
TypeReplacement Part
Unit SizeEach