Eutech™ 10.0 NTU Calibration Standard for TB1000
Product Image
Thermo Scientific™
Eutech™ 10.0 NTU Calibration Standard for TB1000
The CyberScan TB1000 series turbidity meters and its accessories and standards are designed to meet the stringent requirements of today's scientists.
Have Questions?
Catalog NumberVolume (Metric)
01X344318125 mL
Catalog number 01X344318
Price (INR)
-
Request A Quote
Volume (Metric):
125 mL
Eutech™ 10.0 NTU Calibration Standard is used to calibrate the turbidimeter Cyberscan TB1000. The CyberScan TB1000 series turbidity meters and its accessories and standards are designed to meet the stringent requirements of today's scientists.
Features:
  • 125mL bottle
  • Used With: CyberScan TB1000
Specifications
Description10.0 NTU Calibration Standard
Volume (Metric)125 mL
For Use With (Equipment)CyberScan TB 1000
TypeCaibration Standard
Unit SizeEach