DRI™ Ethyl Alcohol Drugs of Abuse Controls
Product Image
Thermo Scientific™

DRI™ Ethyl Alcohol Drugs of Abuse Controls

Eliminate time-consuming steps yet maintain high analytical accuracy with ready-to-use Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Controls in liquid form.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
023950 ng/mL
0243300 ng/mL
Catalog number 0239
Price (INR)
-
Request A Quote
Concentration:
50 ng/mL
Eliminate time-consuming steps yet maintain high analytical accuracy with ready-to-use Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Controls in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration50 ng/mL
DescriptionDRI Alcohol Drugs of Abuse Controls
EthanolEthanol
Quantity1 x 5 mL
TypeControl
Detectable AnalytesAlcohol
Sample TypePlasma, Serum, Urine
Unit SizeEach